Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syndecan 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547377
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547377 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547377 Supplier Invitrogen™ Supplier No. PA547377
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse Syndecan-4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene belongs to the syndecan proteoglycan family. It may play a role in the organization of cell shape by affecting the actin cytoskeleton, possibly by transferring signals from the cell surface in a sugar-dependent mechanism. Allelic variants of this gene have been associated with obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Sdc3
Gene Accession No. Q64519
Gene Alias Kiaa0468; mKIAA0468; Neuroglycan; N-syndecan; SDC3; SDCN; syn-3; Synd3; syndecan 1; syndecan 3; syndecan neural type; syndecan proteoglycan 3; syndecan-3; wu:fa97a01
Gene Symbols Sdc3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-3 Ala45-Glu384.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20970
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.