Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Syndecan 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579961
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human U20S whole cell, rat PC-12 whole cell, mouse NIH/3T3 whole cell. IHC: Human Placenta tissue, Rat Brain tissue.
The protein encoded by this gene belongs to the syndecan proteoglycan family. It may play a role in the organization of cell shape by affecting the actin cytoskeleton, possibly by transferring signals from the cell surface in a sugar-dependent mechanism. Allelic variants of this gene have been associated with obesity.
Specifications
Syndecan 3 | |
Polyclonal | |
Unconjugated | |
Sdc3 | |
Kiaa0468; mKIAA0468; Neuroglycan; N-syndecan; SDC3; SDCN; syn-3; Synd3; syndecan 1; syndecan 3; syndecan neural type; syndecan proteoglycan 3; syndecan-3; wu:fa97a01 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116673, 20970, 9672 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
O75056, P33671, Q64519 | |
Sdc3 | |
A synthetic peptide corresponding to a sequence at the N-terminus of mouse Syndecan 3/SDC3 (45-60aa AQRWRNENFERPVDLE). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction