Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syndecan-4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen Syndecan-4
Concentration 0.5 mg/mL
Dilution Western Blot 1-2 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224630SS
SDP
View Documents
Novus Biologicals
NBP224630SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224630
SDP
View Documents
Novus Biologicals
NBP224630
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Syndecan-4 Polyclonal specifically detects Syndecan-4 in Human, Mouse, Primate samples. It is validated for Western Blot.
Specifications

Specifications

Syndecan-4
Western Blot 1-2 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Primate
amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4
SDC4
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee, 95% homologous in horse, 85% homologous in mouse and rat, and 76% homologous in cow.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Cellular Signaling, Neuroscience
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
6385
A portion of amino acids 1-50 of human Syndecan-4 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.