Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syndecan-4 Antibody (V60P2C2*F2), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250315AF488
Description
Syndecan-4 Monoclonal specifically detects Syndecan-4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Syndecan-4 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SDC4 | |
Peptide Sequence - EDVVGPGQE | |
0.1 mL | |
Cellular Signaling, Neuroscience | |
6385 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
V60P2C2*F2 | |
Western Blot, ELISA, Immunohistochemistry | |
amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction