Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Syndecan 4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595950
Description
Immunogen sequence: ESIRETEVID PQDLLEGRYF SGALPDDEDV VGPGQESDDF ELSGSGDLDD LEDSMIGPEV VHPLVPLDNH IPERAGSGSQ VPTEPKKLEE NEVIPKRISP VEESEDVSNK VSMSSTVQGS NIFERTE; Positive Samples: Mouse kidney; Cellular Location: Membrane, Single-pass type I membrane protein.
Syndecans are type I integral membrane proteoglycans that are involved in cell-extracellular matrix adhesion and growth factor binding. Syndecan 1 is a matrix receptor which binds to Collagens, Fibronectin and Thrombospondin. Syndecan 1 and Syndecan 3 interact with MK (midkine), a growth/differentiation factor involved in embryogenesis. Syndecan 2 is highly expressed at areas of high morphogenetic activity, such as epithelial-mesenchymal interfaces and the prechondrogenic and preosteogenic mesenchymal condensations. Syndecan 4 functions cooperatively with integrins in the processes of cell spreading and focal adhesion assembly.
Specifications
| Syndecan 4 | |
| Polyclonal | |
| Unconjugated | |
| Sdc4 | |
| AA959608; Amphiglycan; AW108331; MGC22217; RATRYUDOCA; RYUDOCA; ryudocan; ryudocan amphiglycan; ryudocan core protein; Ryudocan/syndecan 4; S4; SDC4; Synd4; syndecan 4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; syndecan-4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6385 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P31431 | |
| Sdc4 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction