Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syndecan-4 Rabbit anti-Mouse, Clone: 112, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Syndecan-4 |
---|---|
Clone | 112 |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:25-1:100 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
Syndecan-4 Monoclonal specifically detects Syndecan-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
Syndecan-4 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:25-1:100 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
6385 | |
IgG | |
Protein A purified |
112 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse | |
amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4 | |
Recombinant Mouse Syndecan-4 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title