Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syndecan 4 Recombinant Rabbit Monoclonal Antibody (112), PE
GREENER_CHOICE

Catalog No. PIMA541014
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541014 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541014 Supplier Invitrogen™ Supplier No. MA541014
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

Syndecans are type I integral membrane proteoglycans that are involved in cell-extracellular matrix adhesion and growth factor binding. Syndecan 1 is a matrix receptor which binds to Collagens, Fibronectin and Thrombospondin. Syndecan 1 and Syndecan 3 interact with MK (midkine), a growth/differentiation factor involved in embryogenesis. Syndecan 2 is highly expressed at areas of high morphogenetic activity, such as epithelial-mesenchymal interfaces and the prechondrogenic and preosteogenic mesenchymal condensations. Syndecan 4 functions cooperatively with integrins in the processes of cell spreading and focal adhesion assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan 4
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 112
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene Sdc4
Gene Accession No. O35988
Gene Alias AA959608; Amphiglycan; AW108331; MGC22217; RATRYUDOCA; RYUDOCA; ryudocan; ryudocan amphiglycan; ryudocan core protein; Ryudocan/syndecan 4; S4; SDC4; Synd4; syndecan 4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; syndecan-4
Gene Symbols Sdc4
Host Species Rabbit
Immunogen Recombinant Mouse Syndecan-4/SDC4 protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20971
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.