Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syndetin Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS16108R

Description
CCDC132 is a protein coding gene.Specifications
Syndetin | |
Polyclonal | |
Unconjugated | |
Vps50 | |
1700034M03Rik; 8430415E05Rik; Bcl2-like protein blm; BLM; C87205; Ccdc132; coiled-coil domain containing 132; Coiled-coil domain-containing protein 132; DKFZp313I2429; EARP/GARPII complex subunit VPS50; FLJ20097; KIAA1861; mKIAA1861; RGD1304751; Syndetin; VPS50; VPS50 EARP/GARPII complex subunit; VPS50, EARP/GARPII complex subunit; VPS54L | |
Rabbit | |
Protein A | |
RUO | |
55610 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96JG6 | |
Vps50 | |
KLH conjugated synthetic peptide derived from human Syndetin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction