Learn More
Invitrogen™ SYNDIG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119716
Description
Positive Control: 293 cell lysates, mouse brian tissue, mouse heart tissue, rat brain tissue, rat cerebellum tissue, human skeletal muscletissue, human placenta, F9. Subcellular Location: Cell junction, Cell membrane, Cell projection, Endosome, Membrane, Postsynaptic cell membrane, Synapse.
This gene encodes a protein that belongs to the interferon-induced transmembrane family of proteins. A similar protein in rat is thought to regulate the development of excitatory synapses. [provided by RefSeq, Jul 2013].
Specifications
SYNDIG1 | |
Polyclonal | |
Unconjugated | |
SYNDIG1 | |
AW123113; C20orf39; dispanin subfamily C member 2; DSPC2; FLJ14220; Gm14134; H2C20orf39; IFITMD5; interferon induced transmembrane protein domain containing 5; OTTMUSG00000015750; RGD1310753; synapse differentiation induced gene 1; synapse differentiation inducing 1; synapse differentiation-inducing gene protein 1; SynDIG1; Tmem90b; transmembrane protein 90B | |
Rabbit | |
Affinity chromatography | |
RUO | |
362235, 433485, 79953 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
A2ANU3, Q58DZ9, Q9H7V2 | |
SYNDIG1 | |
Synthetic peptide within human SYNDIG1 aa 100-150. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.