Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYNE1 Antibody (9F10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259946H
Description
SYNE1 Monoclonal specifically detects SYNE1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SYNE1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SYNE1 | |
GST fused to residues 8,165-8,473 of mouse SYNE1 | |
0.1 mL | |
Primary | |
Specificity of human, mouse, rat SYNE1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C in the dark. |
Western Blot | |
9F10 | |
Western Blot | |
ARCA1, C6orf98, CPG2, dJ45H2.2, EC 4.2.1.49, EC 4.3.1.3, EDMD4, Enaptin, KIAA0796chromosome 6 open reading frame 98, KIAA1262FLJ41140, KIAA17568B, Myne-1, MYNE1DKFZp781J13156, Myocyte nuclear envelope protein 1, nesprin 1, nesprin-1, Nuclear envelope spectrin repeat protein 1, SCAR8FLJ30878, spectrin repeat containing, nuclear envelope 1, Synaptic nuclear envelope protein 1, synaptic nuclei expressed gene 1, Syne-1, SYNE-1B | |
Mouse | |
Protein A purified | |
Developmental Biology | |
23345 | |
Human, Mouse, Rat | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction