Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SYNE1, Mouse, Unlabeled, Clone: 7A12,

Catalog No. MABT843 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT843 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT843 Supplier MilliporeSigma™ Supplier No. MABT843
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nesprin-1 (UniProt Q8NF91; also known as Enaptin, Myne-1, Myocyte nuclear envelope protein 1, Nuclear envelope spectrin repeat protein 1, Synaptic nuclear envelope protein 1, Syne-1) is encoded by the SYNE1 (also known as C6orf98, EDMD4, KIAA0796, KIAA1262, KIAA1756, MYNE1, SCAR8) gene (Gene ID 23345) in human. SYNE1 gene encodes multiple intracellular linker/scaffold molecules as a result of alternate start codons usage and splicings. Structurally, nesprins have a central rod domain composed of spectrin repeats. Nesprin-1 isoforms giant/4/5/6/7/10 and nesprin-2 isoforms giant/p32CH/NUANCE-N-33 have two N-terminal CH (calponin homology) domains that bind the actin cytoskeleton. Nesprin-1 isoforms giant/beta/alpha/4/beta 2/alpha 2/myne-1 and Nesprin-2 isoforms giant/2/epsilon2/beta1/alpha1/alpha2/gamma/beta2/FLJ56122/FLJ55476/epsilon1 contain a C-terminal transmembrane KASH (Klarsicht-ANC-Syne-homology) domain that resides in the outer nuclear membrane and bind across the luminal space to the SUN (Sad1, UNC84) type II inner nuclear membrane proteins, SUN1 and SUN2, to form LINC (Linker of Nucleoskeleton and Cytoskeleton) complexes that link the cytoskeleton and the nucleus. Human SYNE1 gene is among the top ten loci conferring risk for the five major psychiatric disorders, schizophrenia, bipolar disorder (BD), major depression, attention deficit-hyperactivity disorder (ADHD), and autism spectrum disorder (ASD), as well as multiple other neurological and neuromuscular disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNE1
Applications Western Blot
Classification Monoclonal
Clone 7A12
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SYNE1, C6orf98, EDMD4, KIAA0796, KIAA1262, KIAA1756, MYNE1, SCAR8
Gene Accession No. Q8NF91
Host Species Mouse
Immunogen GST-tagged recombinant human SYNE1 isoform Beta C-terminal 121-a.a. fragment fused to full-length isoform Alpha sequence (UniProt Q8NF91-2 andQ8NF91-3).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_892006
Test Specificity Epitope region is present in, human isoforms 1 (Giant; Enaptin)/2 (Beta)/3 (Alpha)/4/5/8 (Beta 2)/9 (Alpha 2)/11 (myne-1; 131kDa), mouse isoforms 1 (Giant; Enaptin)/2/3 (Syne-1A)/4 (Syne-1B)), and rat Nesprin-1 (UniProt Q8VHJ9), but absent from human spliced isoforms 6/7/10 (drop1)/12 (GSRP-56; 56 kDa), mouse spliced isoform 5 (Enaptin-165), or rat spliced isoform CPG2 (UniProt Q63128) reported by UniProt (Q8NF91, Q6ZWR6, Q8VHJ9, Q63128). Clone 7A12 does not reacts with nesprin-2, but stained centrosome in acetone/methanol-fixed HeLa cells due to uncharacterized cross-reactivity (Randles, K.N., et al. (2010). Dev. Dyn. 239(3):998-1009).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.