Learn More
SYNE2/Nesprin 2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305393AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
There are two genes encoding members of a new family of type II integral membrane proteins. Both are ubiquitously expressed, and tissue-specific alternative mRNA initiation and splicing generate at least two major isoforms of each protein, with the smaller isoforms being truncated at the N-terminus. These proteins are called nesprin-l and -2 for nuclear _envelope _spectrin repeat, as they are characterized by the presence of multiple, clustered spectrin repeats, bipartite nuclear localization sequences and a conserved C-terminal, single transmembrane domain. Transient transfection of EGFP-fusion expression constructs demonstrated their localization to the nuclear membrane with a novel C-terminal, TM-domain-containing sequence essential for perinuclear localization. Nesprins may function as ′dystrophins of the nucleus′ to maintain nuclear organization and structural integrity.Specifications
SYNE2/Nesprin 2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8WXH0 | |
SYNE2 | |
Between 4200 and 4250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SYNE2 | |
Nuclear envelope spectrin repeat protein 2; Nucleus and actin connecting element protein; Protein NUANCE; Synaptic nuclear envelope protein 2; Syne-2; KIAA1011; NUA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23224 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.