Learn More
Invitrogen™ SYNE3 Recombinant Rabbit Monoclonal Antibody (5P10J3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555501

Description
Immunogen Sequence: LIRNWHLQRM EVDSGKKMVF TNNIPKSGFL INPMDPIPRH RRRANLLQEE EGSHEDFSQL LRNFGQWLQV ENSKLVRIIA MRTSTAEDLR TRKSKLQELE ARVPEGQHLF ENLLRLGPAR GTSDELEDLR YQWMLYKSKL KDSGHLLTQS SPGEPTGFQK TRRWRGLGSL FRRACCVALP LQLLLL.
As a component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. Probable anchoring protein which tethers the nucleus to the cytoskeleton by binding PLEC which can associate with the intermediate filament system. Plays a role in the regulation of aortic epithelial cell morphology, and is required for flow-induced centrosome polarization and directional migration in aortic endothelial cells. [UniProt].
Specifications
SYNE3 | |
Recombinant Monoclonal | |
1.2 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4FZC9, Q6ZMZ3 | |
SYNE3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 750-935 of human Nesprin3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
5P10J3 | |
Unconjugated | |
SYNE3 | |
C14orf139; C14orf49; KASH domain-containing protein 3; KASH3; LINC00341; Nesp3; nesprin-3; NET53; Nuclear envelope spectrin repeat protein 3; RGD1304563; spectrin repeat containing nuclear envelope family member 3; spectrin repeat containing, nuclear envelope family member 3; SYNE3 | |
Rabbit | |
Protein A | |
RUO | |
161176, 212073 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.