Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SYNE4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563827
Description
Immunogen sequence: PRLGSEPLNH PPGAPREADI VGCTVCPASG EESTSPEQAQ TLGQDSLGPP EHFQGGPRGN EPAAHPPRWS TPSSYEDP Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 42%.
The exact function of C19orf46 remains unknown.
Specifications
| SYNE4 | |
| Polyclonal | |
| Unconjugated | |
| SYNE4 | |
| C18H19orf46; C19orf46; C1H19orf46; deafness, autosomal recessive 76; DFNB76; KASH domain-containing protein 4; KASH domain-containing protein C19orf46 homolog; KASH4; Nesp4; Nesprin-4; nuclear envelope spectrin repeat protein 4; RGD1304580; spectrin repeat containing nuclear envelope family member 4; spectrin repeat containing, nuclear envelope family member 4; Syne4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 163183 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N205 | |
| SYNE4 | |
| Recombinant protein corresponding to Human SYNE4. Recombinant protein control fragment (Product #RP-99942). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction