Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SYNE4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542662
Description
Peptide sequence: GEESTSPEQA QTLGQDSLGP PEHFQGGPRG NEPAAHPPRW STPSSYEDPA Sequence homology: Cow: 85%; Horse: 85%; Human: 100%.
The exact function of C19orf46 remains unknown.
Specifications
SYNE4 | |
Polyclonal | |
Unconjugated | |
SYNE4 | |
C18H19orf46; C19orf46; C1H19orf46; deafness, autosomal recessive 76; DFNB76; KASH domain-containing protein 4; KASH domain-containing protein C19orf46 homolog; KASH4; Nesp4; Nesprin-4; nuclear envelope spectrin repeat protein 4; RGD1304580; spectrin repeat containing nuclear envelope family member 4; spectrin repeat containing, nuclear envelope family member 4; Syne4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
163183 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N205 | |
SYNE4 | |
Synthetic peptide directed towards the N-terminal of human C19orf46 (aa 38-87). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction