Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Synechococcus sp. phr Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117626
Description
This antibody has been tested in direct-ELISA.
Involved in repair of UV radiation-induced DNA damage. Catalyzes the light-dependent monomerization (300-600 nm) of cyclobutyl pyrimidine dimers (in cis-syn configuration), which are formed between adjacent bases on the same DNA strand upon exposure to ultraviolet radiation.
Specifications
Synechococcus sp. phr | |
Polyclonal | |
Unconjugated | |
Deoxyribodipyrimidine photo-lyase; Photoreactivating enzyme; phrA | |
Recombinant Synechococcus sp. Deoxyribodipyrimidine photo-lyase protein (2-484aa). | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
4.759 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction