Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SynGAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537910
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537910 Supplier Invitrogen™ Supplier No. PA537910
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with human, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Synaptic Ras-GTPase activating protein (SynGAP) is a PDZ-domain interacting protein expressed almost exclusively in the brain. SynGAP has been shown to be a major component of postsynaptic density and associates with post synaptic density 95 kDa (PSD-95), a synaptic scaffold protein, and synaptic N-methyl-D-aspartate (NMDA) receptors. SynGAP Ras-GTPase activity has also been shown to be inhibited by phosphorylation by CaM kinase II, another abundant protein located in the postsynaptic density. It has been proposed that SynGAP may function as a negative regulator of MAP Kinase activation in the absence of active NMDA receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SynGAP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SYNGAP1
Gene Accession No. F6SEU4, Q96PV0
Gene Alias DKFZp761G1421; Gm1963; KIAA1938; MRD5; Neuronal R; Neuronal RasGAP; OTTHUMP00000209046; p135 SynGAP; Ras GTPase-activating protein SynGAP; ras/Rap GTPase-activating protein SynGAP; RASA1; RASA5; synaptic Ras GTPase activating protein 1; synaptic Ras GTPase activating protein 1 homolog; synaptic Ras GTPase activating protein 1 homolog (rat); synaptic Ras GTPase activating protein, 135kDa; synaptic Ras GTPase-activating protein 1; Synaptic Ras-GAP 1; SYNGAP; Syngap1
Gene Symbols SYNGAP1
Host Species Goat
Immunogen Peptide with sequence C-ESAHIEREEYKLKEY, from the internal region near the C-terminus of SYNGAP1 (aa1169-1183).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240057, 8831
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.