Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SynGAP Recombinant Rabbit Monoclonal Antibody (8M2S9)
GREENER_CHOICE

Catalog No. PIMA542710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542710 Supplier Invitrogen™ Supplier No. MA542710
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Mouse brain, Rat brain. Immunogen sequence: SEKRLRQQQA EKDSQIKSII GRLMLVEEEL RRDHPAMAEP LPEPKKRLLD AQERQLPPLG PTNPRVTLAP PWNGLAPPAP PPPPRLQITE NGEFRNTADH.

Synaptic Ras-GTPase activating protein (SynGAP) is a PDZ-domain interacting protein expressed almost exclusively in the brain. SynGAP has been shown to be a major component of postsynaptic density and associates with post synaptic density 95 kDa (PSD-95), a synaptic scaffold protein, and synaptic N-methyl-D-aspartate (NMDA) receptors. SynGAP Ras-GTPase activity has also been shown to be inhibited by phosphorylation by CaM kinase II, another abundant protein located in the postsynaptic density. It has been proposed that SynGAP may function as a negative regulator of MAP Kinase activation in the absence of active NMDA receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SynGAP
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8M2S9
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SYNGAP1
Gene Accession No. F6SEU4, Q96PV0, Q9QUH6
Gene Alias DKFZp761G1421; Gm1963; KIAA1938; MRD5; Neuronal R; Neuronal RasGAP; OTTHUMP00000209046; p135 SynGAP; Ras GTPase-activating protein SynGAP; ras/Rap GTPase-activating protein SynGAP; RASA1; RASA5; synaptic Ras GTPase activating protein 1; synaptic Ras GTPase activating protein 1 homolog; synaptic Ras GTPase activating protein 1 homolog (rat); synaptic Ras GTPase activating protein, 135kDa; synaptic Ras GTPase-activating protein 1; Synaptic Ras-GAP 1; SYNGAP; Syngap1
Gene Symbols SYNGAP1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192117, 240057, 8831
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.