Learn More
Synoptics™ Syngene SYBR™ Gold Filter
GREENER_CHOICESDP
Catalog No. 01000802 Shop All Synoptics, Inc. Products
Catalog No. 01-000-802 Supplier Synoptics™ Supplier No. FILTSGGX
Add to Cart
Add to Cart
SYBR™ Gold Filter is designed for imaging SYBR Gold™ dyes in GBOX Systems.
SYBR™ Gold Filter
  • Used to image SYBR™ Gold dyes
  • Intended for use in 7 position filter wheel and 3 position filter slider in GBOX Systems
Specifications
Product Line Syngene
For Use With (Application) To Image SYBR™ Gold Dyes
Product Type Filter