Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNGR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589618
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589618 Supplier Invitrogen™ Supplier No. PA589618
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEGGAYGAGK AGGAFDPYTL VRQPHTILRV VSWLFSIVVF GSIVNEGYLN SASEGEEFCI YNRNPNACSY GVAVGVLAFL TCLLYLALDV YFPQISSVKD RKKAVLSDIG VSAFWAFLWF VGFCYLANQW QVSKPKDNPL NEGTDAARAA IAFSFFSIFT WSLTAALAVR RFKDLSFQEE YSTLFPASAQ P; Positive Samples: Mouse pancreas, Rat brain; Cellular Location: Cell junction, Melanosome, Membrane, Multi-pass membrane protein, synapse.

Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear. The gene for SYNGR1is located at chromosome 22q13, a region linked to schizophrenia; however, there is mixed evidence suggesting that mutations in SYNGR1 might be associated with schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNGR1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Syngr1
Gene Accession No. O55100, Q62876
Gene Alias p29; R75468; RP3-333H23.3; synaptogyrin 1; synaptogyrin-1; Syngr1; Syngr1b
Gene Symbols Syngr1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-191 of human SYNGR1 (NP_6637831).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20972, 29205
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.