Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNGR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563698 Supplier Invitrogen™ Supplier No. PA563698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FQDLRNDAPV PYKRFLDEGG MVLTTL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. SYNGR3 is predominantly expressed in brain and placenta. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNGR4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Syngr4
Gene Accession No. O95473
Gene Alias 1700016O14Rik; MGC125805; synaptogyrin 4; synaptogyrin 4 protein; synaptogyrin-4; SYNGR4; testis tissue sperm-binding protein Li 72n
Gene Symbols Syngr4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYNGR4. Recombinant protein control fragment (Product #RP-101729).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23546
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.