Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYNJ2BP/ARIP2 Antibody (OTI3A9), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274417V
Description
SYNJ2BP/ARIP2 Monoclonal specifically detects SYNJ2BP/ARIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
SYNJ2BP/ARIP2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SYNJ2BP | |
Full length recombinant protein of human SYNJ2BP/ARIP2 (NP_060843) produced in HEK293T cells. | |
0.1 mL | |
55333.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI3A9 | |
Western Blot, Flow Cytometry | |
activin receptor interacting protein 5, ARIP2, FLJ11271, Mitochondrial outer membrane protein 25, OMP25FLJ41973, synaptojanin 2 binding protein, synaptojanin-2-binding protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction