Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYNJ2BP/ARIP2 Antibody (OTI3A9), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274417MFV450
Description
SYNJ2BP/ARIP2 Monoclonal antibody specifically detects SYNJ2BP/ARIP2 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
SYNJ2BP/ARIP2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI3A9 | |
Flow Cytometry | |
activin receptor interacting protein 5, ARIP2, FLJ11271, Mitochondrial outer membrane protein 25, OMP25FLJ41973, synaptojanin 2 binding protein, synaptojanin-2-binding protein | |
Full length human recombinant protein of human SYNJ2BP(NP_060843) produced in HEK293T cell. | |
0.1 mL | |
GPCR, Signal Transduction | |
55333 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction