Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNJ2BP Monoclonal Antibody (OTI3A9)
GREENER_CHOICE

Catalog No. PIMA525809
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525809 Supplier Invitrogen™ Supplier No. MA525809
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SYNJ2BP Monoclonal Antibody for Western Blot, Flow

Regulates endocytosis of activin type 2 receptor kinases through the Ral/RALBP1-dependent pathway and may be involved in suppression of activin-induced signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNJ2BP
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3A9
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SYNJ2BP
Gene Accession No. P57105
Gene Alias AA409442; activin receptor interacting protein 2; activin receptor interacting protein 5; activin receptor-interacting protein 2; activin receptor-interacting protein 2a; activin receptor-interacting protein 4; ActRIP4; Arip2; ARIP2a; D12Wsu118e; Mitochondrial outer membrane protein 25; NPW16; Omp25; outer membrane protein; outer membrane protein 25; synaptojanin 2 binding protein; synaptojanin-2-binding protein; SYNJ2BP
Gene Symbols SYNJ2BP
Host Species Mouse
Immunogen Full length human recombinant protein of SYNJ2BP produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55333
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.