Learn More
Invitrogen™ SYNJ2BP Polyclonal Antibody

Description
SYNJ2BP Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
Specifications
Specifications
| Antigen | SYNJ2BP |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 4.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | SYNJ2BP |
| Gene Accession No. | P57105, Q9WVJ4 |
| Gene Alias | AA409442; activin receptor interacting protein 2; activin receptor interacting protein 5; activin receptor-interacting protein 2; activin receptor-interacting protein 2a; activin receptor-interacting protein 4; ActRIP4; Arip2; ARIP2a; D12Wsu118e; Mitochondrial outer membrane protein 25; NPW16; Omp25; outer membrane protein; outer membrane protein 25; synaptojanin 2 binding protein; synaptojanin-2-binding protein; SYNJ2BP |
| Gene Symbols | SYNJ2BP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.