Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SYNM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559053
Description
Immunogen sequence: FLGSGYSSSA TTQQENSYGK AVSSQTNVRT FSPTYGLLRN TEAQVKTFPD RPKAGDTREV PVYIGEDSTI ARESYRDRRD KVAAGASEST RSNE Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 53%.
Type-VI intermediate filament (IF) which plays an important cytoskeletal role within the muscle cell cytoskeleton. It forms heteropolymeric IFs with desmin and/or vimentin, and via its interaction with cytoskeletal proteins alpha-dystrobrevin, dystrophin, talin-1, utrophin and vinculin, is able to link these heteropolymeric IFs to adherens-type junctions, such as to the costameres, neuromuscular junctions, and myotendinous junctions within striated muscle cells.
Specifications
SYNM | |
Polyclonal | |
Unconjugated | |
SYNM | |
4930412K21Rik; AI852401; Desmuslin; DMN; E130104F11; KIAA0353; SYN; synemin; synemin alpha; synemin beta; synemin, intermediate filament protein; synm | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23336 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.40 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15061 | |
SYNM | |
Recombinant protein corresponding to Human SYNM. Recombinant protein control fragment (Product #RP-102883). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction