Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNPO Recombinant Rabbit Monoclonal Antibody (4K5), Proteintech
SDP

Catalog No. 502332690 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-2690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-2690 Supplier Proteintech Group Inc Supplier No. 807211RR100UL
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

SYNPORecombinant Monoclonal antibody specifically detects SYNPO in Human, Mouse, Pig, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SYNPO
Applications Western Blot
Classification Recombinant Monoclonal
Clone 4K5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. Q8N3V7
Gene Alias 9030217H17Rik; 9130229N11; 9330140I15Rik; AW046661; KIAA1029; SYNPO
Host Species Rabbit
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11346
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.