Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNPO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556534 Supplier Invitrogen™ Supplier No. PA556534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDKVGGTPSR EQDAAQTDGL RTTTSYQRKE EESVRTQSSV SKSYIEVSHG LGHVPQQNGF SGASETANIQ RMVPMNRTAK PFPGSVNQPA T Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 69%.

SYNPO2 was initially identified as myopodin, a member of the synaptopodin family that contains one PPXY motif and multiple PXXP motifs. It colocalizes with alpha-actinin and is found at the Z-disc and during stress conditions will translocate to the nucleus, suggesting that it is part of signaling pathways in addition to its function as a structural protein. SYNPO2 has been shown to bind to calmodulin, alpha-actinin, and smooth muscle myosin and will stimulate actin polymerization in a calmodulin dependent manner, consistent with its proposed role in organizing the cytoskeleton. While deletion of SYNPO2 has been reported to be highly correlated with the invasiveness of prostate cancers, other reports suggest that down-regulation of SYNPO2 reduces the invasiveness and motility of prostate cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNPO2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYNPO2
Gene Accession No. Q9UMS6
Gene Alias 1110069I04Rik; 2310068J10Rik; 9530006G20Rik; AI848603; genethonin 2; Genethonin-2; Myo; Myopodin; mypodin; RGD1564779; synaptopodin 2; synaptopodin-2; Synpo2
Gene Symbols SYNPO2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYNPO2. Recombinant protein control fragment (Product #RP-95491).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171024
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.