Learn More
Invitrogen™ SYNPO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588833
Description
Immunogen sequence: MGTGDFICIS MTGGAPWGFR LQGGKEQKQP LQVAKIRNQS KASGSGLCEG DEVVSINGNP CADLTYPEVI KLMESITDSL QMLIKRPSSG ISEALISENE NKNLEHLTHG GYVESTTLQI RPATKTQCTE FFLAPVKTEV PLAENQRSGP DCAGSLKEET GPSYQRAPQM PDSQRGRVAE ELILREKVEA VQPGPVVELQ LSLSQERHKG ASGPLVALPG AEKSKSPDPD PNLSHDRIVH INSIPTNEKA DPFLRSSKII QISSGRELRV IQESEAGDAG LPRVEVILDC SDRQKTEGCR; Positive Samples: Mouse skeletal muscle; Cellular Location: Cytoplasm, Nucleus.
SYNPO2 was initially identified as myopodin, a member of the synaptopodin family that contains one PPXY motif and multiple PXXP motifs. It colocalizes with alpha-actinin and is found at the Z-disc and during stress conditions will translocate to the nucleus, suggesting that it is part of signaling pathways in addition to its function as a structural protein. SYNPO2 has been shown to bind to calmodulin, alpha-actinin, and smooth muscle myosin and will stimulate actin polymerization in a calmodulin dependent manner, consistent with its proposed role in organizing the cytoskeleton. While deletion of SYNPO2 has been reported to be highly correlated with the invasiveness of prostate cancers, other reports suggest that down-regulation of SYNPO2 reduces the invasiveness and motility of prostate cancer cells.
Specifications
SYNPO2 | |
Polyclonal | |
Unconjugated | |
SYNPO2 | |
1110069I04Rik; 2310068J10Rik; 9530006G20Rik; AI848603; genethonin 2; Genethonin-2; Myo; Myopodin; mypodin; RGD1564779; synaptopodin 2; synaptopodin-2; Synpo2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
118449 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.09 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q91YE8 | |
SYNPO2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SYNPO2 (NP_5977342). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.