Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNPO2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563082 Supplier Invitrogen™ Supplier No. PA563082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAEALLLPHG PLRPGPHLIP MVGPVPHPVA EDLTTTYTQK AKQAKLQRAE SLQEKSIKEA KTKCRTIASL LTA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.

SYNPO2L was initially identified as a novel heart-enriched gene that encodes a cytoskeletal protein highly expressed in the Z-disc of heart and skeletal muscle, associates with actin and interacts with a-actinin. It is a member of the synaptopodin family, sharing greatest homology with Synaptopodin 2. Recent studies have shown that SYNPO2L, while primarily localized to the sarcomere, can also translocate to the nucleus. A knockdown of SYNPO2L in zebrafish resulted in aberrant cardiac and skeletal muscle development and function, suggesting that it is a critical component of the sarcomere and plays an important role in muscle development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNPO2L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYNPO2L
Gene Accession No. Q9H987
Gene Alias 1110054M18Rik; Chap; RGD1565434; synaptopodin 2 like; synaptopodin 2-like; synaptopodin 2-like protein; SYNPO2L
Gene Symbols SYNPO2L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYNPO2L. Recombinant protein control fragment (Product #RP-101447).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79933
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.