Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYNRG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570724 Supplier Invitrogen™ Supplier No. PA570724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

May play a role in endocytosis and/or membrane trafficking at the trans-Golgi network (TGN). May act by linking the adapter protein complex AP-1 to other proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNRG
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SYNRG
Gene Accession No. Q9UMZ2
Gene Alias adaptor-related protein complex 1 gamma subunit-binding protein 1; AP1 gamma subunit binding protein 1; AP1 subunit gamma-binding protein 1; Ap1gbp1; fj67h08; Gamma-synergin; MGC104959; synergin gamma; synergin, gamma; Syng; Synrg; wu:fj67h08
Gene Symbols SYNRG
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human SYNRG.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11276
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.