Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntabulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548084
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548084 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548084 Supplier Invitrogen™ Supplier No. PA548084
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Syntabulin/GOLSYN is part of a kinesin motor-adaptor complex that is critical for the anterograde axonal transport of active zone components and contributes to activity-dependent presynaptic assembly during neuronal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntabulin
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SYBU
Gene Accession No. Q9NX95
Gene Alias FLJ20366; Golgi-localized protein; golgi-localized syntaphilin-related protein; Golsyn; GOLSYN A protein; GOLSYN B protein; GOLSYN C protein; Golsyn/Syntabulin; implicated in syntaxin trafficking in neurons; KIAA1472; m-Golsyn; microtubule-associated protein; OCSYN; RGD1564335; SNPHL; SYBU; Syntabulin; syntabulin (syntaxin-interacting); syntaxin-1-binding protein
Gene Symbols SYBU
Host Species Goat
Immunogen E. coli-derived recombinant human Syntabulin Gln489-Arg656.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55638
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.