Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntabulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562721 Supplier Invitrogen™ Supplier No. PA562721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STPGANVLEL LPIVMGQEEG SVVVERAVQT DVVPYSPAIS ELIQSVLQKL QDPCPSSLAS PDESEPDSME SFPESLSALV VDLTPRNPNS AI Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.

Syntabulin/GOLSYN is part of a kinesin motor-adaptor complex that is critical for the anterograde axonal transport of active zone components and contributes to activity-dependent presynaptic assembly during neuronal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntabulin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYBU
Gene Accession No. Q9NX95
Gene Alias FLJ20366; Golgi-localized protein; golgi-localized syntaphilin-related protein; Golsyn; GOLSYN A protein; GOLSYN B protein; GOLSYN C protein; Golsyn/Syntabulin; implicated in syntaxin trafficking in neurons; KIAA1472; m-Golsyn; microtubule-associated protein; OCSYN; RGD1564335; SNPHL; SYBU; Syntabulin; syntabulin (syntaxin-interacting); syntaxin-1-binding protein
Gene Symbols SYBU
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYBU. Recombinant protein control fragment (Product #RP-93815).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55638
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.