Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntaphilin Antibody (M371), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323428
Description
Syntaphilin Monoclonal antibody specifically detects Syntaphilin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Syntaphilin | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
M371 | |
Western Blot 1:500 | |
bA314N13.5, KIAA0374, MGC46096, syntaphilin | |
Clone M371 was generated from a sequence corresponding to amino acids in the N-terminal region of human syntaphilin. This sequence has high homology to mouse and rat syntaphilin. [UniProt# O15079] | |
0.1 mL | |
Neuroscience | |
9751 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction