Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntaphilin Monoclonal Antibody (M371)
GREENER_CHOICE

Catalog No. PIMA547739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547739 Supplier Invitrogen™ Supplier No. MA547739
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody detects a 70 kDa* protein corresponding to the molecular mass of Syntaphilin on SDS-PAGE immunoblots of adult mouse brain.

Syntaphilin was initially identified in a yeast two-hybrid screen with the carboxy terminal region of Syntaxin-1 as bait. Syntaxin-1 is a key component of the synaptic vesicle docking machinery that forms the SNARE complex with synaptobrevin and SNAP-25. Syntaphilin competes with SNAP-25 for binding to syntaxin-1 and inhibits the formation of the SNARE complex, thereby potentially regulating synaptic vesicle exocytosis. Syntaphilin also binds dynamin-1 and inhibits dynamin-dependent endocytosis. Mice lacking syntaphilin show an increased level of mitochondrial motility and a reduced density of axonal mitochondria. This correlates with an enhanced short-term facilitation and significant impairments in motor ability, suggesting syntaphilin plays a major role in presynaptic function. Multiple isoforms are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaphilin
Applications Western Blot
Classification Monoclonal
Clone M371
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene SNPH
Gene Accession No. B5DF41, O15079, Q80U23
Gene Alias 6430515A01; AW045671; AW556958; AW743098; bA314N13.5; KIAA0374; mKIAA0374; Snph; Syntaphilin
Gene Symbols SNPH
Host Species Mouse
Immunogen Clone M371 was generated from a sequence corresponding to amino acids in the N-terminal region of human syntaphilin. This sequence has high homology to mouse and rat syntaphilin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241727, 296267, 9751
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.