Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntaphilin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588474 Supplier Invitrogen™ Supplier No. PA588474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEAGVQASCM QERAIQTDFV QYQPDLDTIL EKVTQAQVCG TDPESGDRCP ELDAHPSGPR DPNSAVVVTV GDELEAPEPI TRGPTPQRPG ANPNPGQSVS VVCPMEEEEE AAVAEKEPKS YWSRHY; Positive Samples: Mouse intestine, Rat brain, Rat testis; Cellular Location: Cell junction, Membrane, Single-pass membrane protein, synapse, synaptosome.

Syntaphilin was initially identified in a yeast two-hybrid screen with the carboxy terminal region of Syntaxin-1 as bait. Syntaxin-1 is a key component of the synaptic vesicle docking machinery that forms the SNARE complex with synaptobrevin and SNAP-25. Syntaphilin competes with SNAP-25 for binding to syntaxin-1 and inhibits the formation of the SNARE complex, thereby potentially regulating synaptic vesicle exocytosis. Syntaphilin also binds dynamin-1 and inhibits dynamin-dependent endocytosis. Mice lacking syntaphilin show an increased level of mitochondrial motility and a reduced density of axonal mitochondria. This correlates with an enhanced short-term facilitation and significant impairments in motor ability, suggesting syntaphilin plays a major role in presynaptic function. Multiple isoforms are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaphilin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.889 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SNPH
Gene Accession No. B5DF41, Q80U23
Gene Alias 6430515A01; AW045671; AW556958; AW743098; bA314N13.5; KIAA0374; mKIAA0374; Snph; Syntaphilin
Gene Symbols SNPH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-425 of human SNPH (NP_0555382).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241727, 296267
Target Species Mouse, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.