Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntaxin 1 Monoclonal Antibody (SP6), Biotin
GREENER_CHOICE

Catalog No. PIMA517613
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517613 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517613 Supplier Invitrogen™ Supplier No. MA517613
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17613 targets Syntaxin-1 in ELISA, FACS, and WB applications and shows reactivity with Human, Mouse, and Rat samples. The MA5-17613 immunogen is human Syntaxin.

Syntaxin 1 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE [Soluble NSF (N-ethylmaleimide-sensitive fusion protein)-Attachment protein REceptor] domain (known as H3), and an N-terminal regulatory domain (Habc). Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone SP6
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Stx1a
Gene Accession No. O35526, P32851, Q16623
Gene Alias HPC-1; LOW QUALITY PROTEIN: syntaxin-1A; neuron-specific antigen HPC-1; P35-1; P35A; Sap; STX1; Stx1a; STX1B; STX1B1; STX1B2; STXA1; SYN1A; Synaptotagmin-associated 35 kDa protein; syntaxin 1 a; syntaxin 1A; syntaxin 1A (brain); syntaxin 1A (brain)-like; syntaxin-1A; Syntaxin-1B1; Syntaxin-1B2
Gene Symbols Stx1a
Host Species Mouse
Immunogen Human Syntaxin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116470, 20907, 6804
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.