Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntaxin 12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186985
Description
Syntaxin 12 Polyclonal specifically detects Syntaxin 12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Syntaxin 12 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
MGC51957, STX13, STX14, syntaxin 12, syntaxin-12 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of human Syntaxin 12 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
STX12 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MSYGPLDVYRNPGPSGPQLRDFSSIIQTCSGNIQRISQATAQIKNLMSQLGTKQDSSKLQENLQQLQHSTNQLAKETNELLKE | |
0.1 mL | |
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
23673 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction