Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 1A / Syntaxin 1B Mouse anti-Human, Mouse, Rat, Clone: 2D3B5, Proteintech
SDP

Catalog No. 501736863 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6863 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6863 Supplier Proteintech Group Inc Supplier No. 664371IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE domain, and an N-terminal regulatory domain. Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Syntaxin 1A / Syntaxin 1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2D3B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STX1A
Gene Accession No. O35526, P32851, P61264, P61265, P61266, Q16623
Gene Alias STX1B, STX1B1, STX1B2, syntaxin 1B, Syntaxin 1B1, Syntaxin 1B2
Gene Symbols STX1A, Stx1b
Host Species Mouse
Immunogen Syntaxin 1A / Syntaxin 1B Fusion Protein Ag10257
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112755, 116470, 20907, 24923, 56216, 6804
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.