Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntaxin 1B Antibody (SR1555), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Syntaxin 1B |
---|---|
Clone | SR1555 |
Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Syntaxin 1B Monoclonal antibody specifically detects Syntaxin 1B in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Syntaxin 1B | |
Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
112755 | |
IgG | |
Affinity purified |
SR1555 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
FP17469, STX1B1, STX1B2, syntaxin 1B, syntaxin 1B1, syntaxin 1B2, syntaxin-1B, syntaxin-1B1, syntaxin-1B2 | |
A synthesized peptide derived from human Syntaxin 1B (Uniprot #: P61266) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title