Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntaxin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA125502

Catalog No. PIPA125502


Only null left
Add to Cart

Description

Description

Recommended positive controls: HeLa Cell Lysate, Rat Brain Tissue Extract, Mouse Brain Tissue Extract.

The product of this gene belongs to the syntaxin/epimorphin family of proteins. The syntaxins are a large protein family implicated in the targeting and fusion of intracellular transport vesicles. The product of this gene regulates epithelial-mesenchymal interactions and epithelial cell morphogenesis and activation. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Syntaxin 2
Polyclonal
Unconjugated
Stx2
AW538950; EPIM; Epimorphin; EPM; G1-536-1; I79_024396; MGC51014; repro34; Stx2; STX2A; STX2B; STX2C; Syn-2; syntaxin 2; syntaxin-2
Rabbit
Antigen affinity chromatography
RUO
100155887, 100352556, 100768784, 101118731, 13852, 2054, 25130
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.2
P32856, P50279, Q00262
Stx2
Synthetic peptide, conjugated to KLH, corresponding to the N-terminal residues M(1) R D R L P D L T A C R K S D D G D N(19) of rat Syntaxin 2.
50 μg
Primary
Human, Mouse, Rat, Rabbit, Monkey, Sheep, Hamster, Pig, Drosophila
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.