Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntaxin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587903 Supplier Invitrogen™ Supplier No. PA587903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRDRLPDLTA CRKNDDGDTV VVVEKDHFMD DFFHQVEEIR NSIDKITQYV EEVKKNHSII LSAPNPEGKI KEELEDLNKE IKKTANKIRA KLKAIEQSFD QDESGNRTSV DLRIRRTQHS VLSRKFVEAM AEYNEAQTLF RERSKGRIQR QLEITGRTTT DDELEEMLES GKPSIFTSDI ISDSQITRQA LNEIESRHKD IMKLETSIRE LHEMFMDMAM FVETQGEMIN NIERNVMNAT DYVEHAKEET KKAIKYQSKA RRKL; Positive Samples: K-562, DU145, SKOV3, A-549, Mouse heart, Mouse testis, Rat liver, Rat heart, Rat testis; Cellular Location: Membrane, Single-pass type IV membrane protein.

The product of this gene belongs to the syntaxin/epimorphin family of proteins. The syntaxins are a large protein family implicated in the targeting and fusion of intracellular transport vesicles. The product of this gene regulates epithelial-mesenchymal interactions and epithelial cell morphogenesis and activation. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Stx2
Gene Accession No. P32856, P50279, Q00262
Gene Alias AW538950; EPIM; Epimorphin; EPM; G1-536-1; I79_024396; MGC51014; repro34; Stx2; STX2A; STX2B; STX2C; Syn-2; syntaxin 2; syntaxin-2
Gene Symbols Stx2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-264 of human STX2 (NP_0019712).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13852, 2054, 25130
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.