Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin-3B Mouse anti-Human, Mouse, Bovine, Clone: 12E5, MilliporeSigma™

Catalog No. MABN2394MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2394MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2394MI Supplier MilliporeSigma Supplier No. MABN2394
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Syntaxin-3B clone 12E5 is a highly specific mouse monoclonal antibody that targets Syntaxin 3B and has been tested in Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin-3B
Applications Immunocytochemistry, Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 12E5
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Stx3
Host Species Mouse
Immunogen GST-tagged recombinant murine Syntaxin 3, isoform B that lacks the transmembrane domain.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001020478
Test Specificity Clone 12E5 is a highly specific antibody against the B isoform of Syntaxin 3, which is abundantly expressed in photoreceptors of the retina.
Target Species Human, Mouse, Bovine
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.