Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntaxin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551560 Supplier Invitrogen™ Supplier No. PA551560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRDRTHELRQ GDDSSDEEDK ERVALVVHPG TARLGSPDEE FFHKVRTIRQ TIVKLGNKVQ ELEKQQVTIL ATPLPEESMK QELQNLRDEI KQLGREIRLQ LKAIEPQKEE ADENYNSVNT RMRKTQHGVL SQQFVELINK CNSMQSE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

STX4 is a plasma membrane t-SNARE that mediates docking of transport vesicles.STX4 is necessary for the translocation of SLC2A4 from intracellular vesicles to the plasma membrane. Together with STXB3 and VAMP2, STX4 may also play a role in docking/fusion of intracellular GLUT4-containing vesicles with the cell surface in adipocytes. STX4 may also play a role in docking of synaptic vesicles at presynaptic active zones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STX4
Gene Accession No. Q12846
Gene Alias hypothetical protein LOC508675; p35-2; Renal carcinoma antigen NY-REN-31; Stx4; STX4A; Syn4; Syn-4; syntaxin 4; syntaxin 4A (placental); syntaxin-4; syntaxin-4 isoform A
Gene Symbols STX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STX4. Recombinant protein control fragment (Product #RP-95939).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6810
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.