Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syntaxin 6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583429 Supplier Invitrogen™ Supplier No. PA583429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFNLDATELS IRKAFITSTR QVVRDMKDQM STSSVQALAE RKNRQALLGD SGSQNWSTGT TDKYGRLDRE LQRANSHFIE EQQAQ Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 88%.

Syntaxin 6 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE domain, and an N-terminal regulatory domain. Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. Syntaxin is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STX6
Gene Accession No. O43752
Gene Alias 2310039E05Rik; 2410005I16Rik; AA437559; AI415714; AI845861; HGNC:11441; STX6; Syn6; syntaxin 6; syntaxin-6
Gene Symbols STX6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STX6. Recombinant protein control fragment (Product #RP-97610).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10228
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.