Learn More
Syntaxin 6 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304397AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
STX6 (Syntaxin 6) is a Protein Coding gene. Diseases associated with STX6 include Supranuclear Palsy, Progressive, 1. Among its related pathways are Delta508-CFTR traffic/ER-to-Golgi in CF and Vesicle-mediated transport. Gene Ontology (GO) annotations related to this gene include syntaxin binding and SNAP receptor activity.Specifications
Syntaxin 6 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O43752, Q9JKK1 | |
STX6 | |
Between 75 and 125 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
STX6 | |
Syntaxin-6, Stx6, Syntaxin 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10228, 58244 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.