Learn More
Syntaxin 7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304512AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
May be involved in protein trafficking from the plasma membrane to the early endosome (EE) as well as in homotypic fusion of endocytic organelles. Mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes.Specifications
Syntaxin 7 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O15400, O70439 | |
STX7 | |
Between 100 and 150 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
STX7 | |
Syntaxin-7, STX7, Syn7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53331, 8417 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.