Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntaxin 8 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298825AF532
Description
Syntaxin 8 Polyclonal specifically detects Syntaxin 8 in Human samples. It is validated for Western Blot.Specifications
| Syntaxin 8 | |
| Polyclonal | |
| Western Blot | |
| CARB, CIP-1-associated regulator of cyclin B, syntaxin 8, syntaxin-8 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Syntaxin 8. | |
| 0.1 mL | |
| Endosome Markers | |
| 9482 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction