Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntaxin 8 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300260PE
Description
Syntaxin 8 Polyclonal antibody specifically detects Syntaxin 8 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Syntaxin 8 | |
Polyclonal | |
ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
CARB, CIP-1-associated regulator of cyclin B, syntaxin 8, syntaxin-8 | |
Produced in rabbits immunized with purified, recombinant Human Syntaxin 8 (Uniprot#: Q9UNK0; Met1-Gly215) | |
0.1 mL | |
Endosome Markers | |
9482 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction