Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntenin 1 Antibody (19C1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Syntenin 1 |
---|---|
Clone | 19C1 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Syntenin 1 Monoclonal antibody specifically detects Syntenin 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
Syntenin 1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MDA9, MDA-9, melanoma differentiation associated protein-9, Melanoma differentiation-associated protein 9, Pro-TGF-alpha cytoplasmic domain-interacting protein 18, Scaffold protein Pbp1, ST1, SYCLTACIP18, syndecan binding protein (syntenin), Syndecan-binding protein 1, syntenin-1 | |
A synthesized peptide derived from Human Syntenin 1 [UniProt O00560] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
19C1 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytokines and Growth Factors, Immunology, Neuroscience | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6386 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title